Learn More
Hardy Diagnostics™ AnaeroGRO Chopped Meat Glucose Broth, Tube with Needle-Port Hungate Screw-Cap
SDP
Catalog No. 21500105 Shop All Hardy Diagnostics Products
Catalog No. 21-500-105 Supplier Hardy Diagnostics™ Supplier No. AG19H
Add to Cart
Add to Cart
AnaeroGRO™ Chopped Meat Media is recommended for the cultivation of aerobic, microaerophilic, and anaerobic microorganisms, especially Clostridium species.
Flat bottom
  • Contributes to stability when set on end
Leak-proof
  • Caps provide security during handling and transportation
Optically clear
  • Tubes are made of clarified glass for clear viewing
Needleport Septum
  • To avoid oxygen exposure, liquid specimens may be injected directly through the rubber septum of the hungate screw cap with a needle and syringe
Specifications
Manufacturing Origin United States
Product Type Tubed Media
Sterility Non-sterile
Target Organism Class Gram-negative, Rod-shaped Anaerobes, Anaerobes
Test Type Microbial Detection, Media Testing, Manual Diagnostics, Growth Promotion Testing
Dimensions (Dia. x H) 16 x 125 mm