Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ AnaeroGRO™ MultiPak A
SDP

Catalog No. 23001740 Shop All Hardy Diagnostics Products
Change view
Click to view available options
No. per Pack:
3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
23-001-740 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-740 Supplier Hardy Diagnostics™ Supplier No. AG303
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This MuliPak comes with 3 different plates: Brucella with Hemin and Vitamin K plate, PEA plate, and BBE/LKV Biplate

  • AnaeroGro™ Brucella Agar with Hemin and Vitamin K is recommended for use in the primary isolation, quantitation, and partial identification of obligately anaerobic microorganisms from clinical specimens. The medium is also suitable for the growth of aerobic and microaerophilic bacteria when incubated under the appropriate conditions.
  • Anaerobic PEA (Phenylethyl Alcohol) Agar is recommended for use as an enriched and selective medium for the cultivation and selective isolation of gram-positive and negative obligate anaerobic bacteria. It is useful in isolating obligate anaerobes from mixed flora, by inhibiting gram-negative facultative anaerobes and the control of swarming organisms.
  • Bacteroides Bile Esculin Agar is an enriched, selective, and differential medium recommended for the rapid isolation and presumptive identification of obligately anaerobic gram-negative bacilli of the Bacteroides fragilis group Bilophila spp.
  • LKV (Laked Blood with Kanamycin and Vancomycin) Agar is recommended for use in the selective isolation and partial identification of obligately anaerobic gram-negative bacilli, such as Prevotella spp. and Bacteroides spp.
  • Features and Benefits:
    • Pre-reducted culture media packaged in oxygen-free gas flushed foil pouches
    • Packaging contains an oxygen scavenger sachet
    • Packaging contains a moisture absorbing desiccant packet
    • Room temperature storage
    • Foil pouches have an easy-to-open pre-scored notch
    • Manufactured in an ISO 13485 certified facility
    • Wide variety of packaging combinations available: monoplates, biplates, and primary set-up combinations

Specifications

Appearance Variable
Certifications/Compliance FDA, ISO 13485, ISO 17025
Country of Origin United States
Additive Hemin, Vitamin K, Bile, Esculin, Kanamycin, Vancomycin, Phenylethyl Alcohol
Detects Bacteroides fragilis Group, Bilophila Spp., Prevotella Spp., Bacteroides Spp.
Form Agar
Format Plate
Manufacturing Origin United States
Product Type Agar Plate
Sterility Sterile
Target Organism Class Gram-negative Bacteria, Anaerobes, Gram-positive Organisms, Gram-negative Bacilli
Test Type Microbial Identification
Fill Volume 17 mL
For Use With (Application) Isolation, Quantitation, Identification of Obligately Anaerobic Microorganisms; Cultivation and Selective Isolation of Gram-Positive and Negative Obligate Anaerobic Bacteria
Incubation Conditions 35°C to 37°C, Anaerobic
Incubation Time 18 to 24 hr.
No. per Pack 3
Quantity 3 Plates/Pouch
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Bacteroides species, Prevotella species
Manufacturing Quality FDA Licensed Facility, ISO 13485, ISO 17025
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.