Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ AnaeroGRO™ MultiPak B
SDP

Catalog No. 23001777 Shop All Hardy Diagnostics Products
Change view
Click to view available options
No. per Pack:
3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
23-001-777 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-777 Supplier Hardy Diagnostics™ Supplier No. AG313
Add to Cart
Add to Cart
This item is not returnable. View return policy

This MultiPak comes with three plates: Brucella with Hemin and Vitamin K plate, LVK plate, and PEA plate

  • AnaeroGro™ Brucella Agar with Hemin and Vitamin K is recommended for use in the primary isolation, quantitation, and partial identification of obligately anaerobic microorganisms from clinical specimens. The medium is also suitable for the growth of aerobic and microaerophilic bacteria when incubated under the appropriate conditions.
  • Anaerobic PEA (Phenylethyl Alcohol) Agar is recommended for use as an enriched and selective medium for the cultivation and selective isolation of gram-positive and negative obligate anaerobic bacteria. It is useful in isolating obligate anaerobes from mixed flora, by inhibiting gram-negative facultative anaerobes and the control of swarming organisms.
  • AnaeroGRO™ LKV (Laked Blood with Kanamycin and Vancomycin) Agar is recommended for use in the selective isolation and partial identification of obligately anaerobic gram-negative bacilli, such as Prevotella spp. and Bacteroides spp.
  • Features and Benefits:
    • Pre-reducted culture media packaged in oxygen-free gas flushed foil pouches
    • Packaging contains an oxygen scavenger sachet
    • Packaging contains a moisture absorbing desiccant packet
    • Room temperature storage
    • Foil pouches have an easy-to-open pre-scored notch
    • Manufactured in an ISO 13485 certified facility
    • Wide variety of packaging combinations available: monoplates, biplates, and primary set-up combinations

Specifications

Certifications/Compliance FDA
Country of Origin United States
Additive Hemin, Vitamin K, Kanamycin, Vancomycin
Detects Prevotella spp. and Bacteroides spp.
Form Prepared Medium
Format Plate
Manufacturing Origin United States
Media Type Brucella with H and K, LVK, and PEA
Product Type Agar Plate
Sterility Sterile
Target Organism Class Anaerobes, Gram-negative Bacilli
Test Type Microbial Identification
Fill Volume 17 mL
For Use With (Application) Primary isolation, quantitation, and partial identification of obligately anaerobic microorganisms from clinical specimens, Growth of aerobic and microaerophilic bacteria, Cultivation and selective isolation of Gram-positive and negative obligate anaerobic bacteria, Selective isolation and partial identification of obligately anaerobic Gram-negative bacilli
Material Polystyrene
No. of Places 3
No. per Pack 3
Quantity 3 Plates/Pouch
Diameter (Metric) 100 mm
Species Bacteroides species, Porphyromonas species, Clostridium species, Prevotella species
Manufacturing Quality FDA Licensed Facility
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.