Description
Mitsubishi™ AnaeroPack-CO2 is used for cultivation of CO2 dependent microorganisms such as Haemophilus and Neisseria spp.
- Use 1 sachet/2.5L jar or 3 sachets/7.0L jar
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Type | AnaeroPack-CO2 |
| Format | Gas Generation Sachet |
| Product Line | Thermo Scientific Mitsubishi |
| Quantity | 20/Pk. |