Learn More
Thermo Scientific™ AnaeroPouch™-Anaero Anaerobic Gas Generator
GSA_VA
Catalog No. 23246379 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20/Pk.
Catalog No. Quantity
23-246-379 20/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 23-246-379 Supplier Thermo Scientific™ Supplier No. R682001
Add to Cart
Add to Cart
This item is not returnable. View return policy
Mitsubishi™ AnaeroPouch-Anaero is used for cultivation of anaerobic microorganisms without the addition of water or use of a catalyst.
Mitsubishi™ AnaeroPouch-Anaero is used for cultivation of anaerobic microorganisms without the addition of water or use of a catalyst.
  • No hydrogen gas produced
  • Use 1 sachet/AnaeroPouch-Bag or 0.4L jar with 1 to 2 plates
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Type AnaeroPouch-Anaero
Format Gas Generation Sachet
Product Line Thermo Scientific Mitsubishi
Quantity 20/Pk.