Learn More
Thermo Scientific™ AnaeroPouch™-CO2 Generator
GSA_VA
Catalog No. 23246381 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20/Pk.
Catalog No. Quantity
23-246-381 20/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 23-246-381 Supplier Thermo Scientific™ Supplier No. R682007
Add to Cart
Add to Cart
This item is not returnable. View return policy
Mitsubishi™ AnaeroPouch-CO2 is used for cultivation of CO2 dependent microorganisms
Mitsubishi™ AnaeroPouch-CO2 is used for cultivation of CO2 dependent microorganisms such as Haemophilus and Neisseria spp.
  • Use 1 sachet/AnaeroPouch-Bag or 0.4L jar with 2 plates
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Type AnaeroPouch-CO2
Format Gas Generation Sachet
Product Line Thermo Scientific Mitsubishi
Quantity 20/Pk.