Learn More
Thermo Scientific™ AnaeroPouch™-Keep System
Catalog No. 23246382 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20/Pk.
Catalog No. Quantity
23-246-382 20/Pk.
1 options
Catalog No. 23-246-382 Supplier Thermo Scientific™ Supplier No. R682015
Add to Cart
Add to Cart
Reduce prepared culture media to less than 0.1% oxygen in 8 hours with an oxygen-absorbing system for use in cultivation of anaerobic bacteria.
Reduce prepared culture media to less than 0.1% oxygen in 8 hours with Mitsubishi™ AnaeroPouch-Keep, an oxygen-absorbing system, for use in cultivation of anaerobic bacteria.
  • Use one AnaeroPouch-Keep sachet/AnaeroPouch-Bag, purchased separately Part No. R686001
  • Up to six plates can be reduced in each AnaeroPouch-Bag
TRUSTED_SUSTAINABILITY
Specifications
Type AnaeroPouch-Keep
Format Pouch
Product Line Thermo Scientific Mitsubishi
Quantity 20/Pk.