Learn More
Medchemexpress LLC HY-14877 5mg Medchemexpress, Anagliptin CAS:739366-20-2 Purity:>98%
SDP
Catalog No. 502030563
Encompass_Preferred
Medchemexpress, HY-14877 5mg Anagliptin CAS:739366-20-2 Anagliptin is a highly selective, potent inhibitor of dipeptidyl peptidase 4 (DPP-4), with an IC50 of 3.8 nM, and less selective at DPP-8/9 (IC50, 68, 60 nM, respectively). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0563 Supplier Medchemexpress LLC Supplier No. HY148775MG
Medchemexpress, HY-14877 5mg Anagliptin CAS:739366-20-2 Anagliptin is a highly selective, potent inhibitor of dipeptidyl peptidase 4 (DPP-4), with an IC50 of 3.8 nM, and less selective at DPP-8/9 (IC50, 68, 60 nM, respectively). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.