Learn More
Electron Microscopy Sciences Analine Eosin Solution 250 ML
SDP
Catalog No. 5031663
Encompass_Preferred
Used in Gridley's Method for Endamoeba Histolytica. For bacteria stain: Amoebae, their nuclei and ingested erythrocytes.
Catalog No. 50-316-63 Supplier Electron Microscopy Sciences Supplier No. 2610902
Used in Gridley's Method for Endamoeba Histolytica. For bacteria stain: Amoebae, their nuclei and ingested erythrocytes.