Learn More
Vernier™ Breadboard Cable (Analog)
SDPSCI_ED
Catalog No. S16082ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Interfaces
Catalog No. For Use With (Equipment)
S16082ND Interfaces
1 options
Catalog No. S16082ND Supplier Vernier™ Supplier No. BBBTA
Add to Cart
Add to Cart
Includes
Breadboard Cable (Analog)
The Breadboard Cable (Analog) provides an easy way for students to build their own sensor circuitry and input the signal into a Vernier interface for data acquisition.
Pin Information There are six pins available: SIG1, 5V, ID, VRES, GND, and SIG2. See full specifications and troubleshooting guide »
Warranty and Services
5-year limited warranty
Specifications
Product Type Analog
For Use With (Equipment) Interfaces
Description Six pins available: SIG1, 5V, ID, VRES, GND, and SIG2
For Use With (Application) Educational use only