Learn More
Vernier™ Analog BTA Cable
SDPSCI_ED
Catalog No. S16115ND Shop All Vernier Software Products
Change view
Click to view available options
Length (Metric):
1.5 m
Catalog No. Length (Metric)
S16115ND 1.5 m
1 options
Catalog No. S16115ND Supplier Vernier™ Supplier No. CBBTA
Add to Cart
Add to Cart
Includes
Analog BTA Cable
This cable has bare wire on one end and a male BTA connector on the other end. This cable can be used to connect your own sensors to our interfaces. The cable is approximately 5 feet long.
  • This cable can be used to connect your homemade analog sensors you make to interfaces.
  • There is a BTA plug on on end and six bare wires on the other end.
  • Use a multimeter and test for continuity to determine which wire goes with which pin of the cable's BTA connector.
Warranty and Services
1 year
Specifications
For Use With (Equipment) Interfaces
Length (Metric) 1.5 m
For Use With (Application) Educational use only