Learn More
Research Products International Corp Analog HotPlate and Stirrer
SDP
Catalog No. 502229786
Encompass
Popular HotPlate and Stirrer features a durable, chemically resistant white ceramic plate. Ideal space efficient design allows use on crowded benchtops or inside of hoods. Advanced microprocessor controls feature convenient turn knobs for quick, precise adjustment of speed and temperature.
LED light on front panel visually indicates when heat and/or stirring functions are activated. Work surface of 7 1/2 inches allows for a variety or laboratory vessels. Supplied with support rod for mounting thermometer or temperature probe.
Catalog No. 50-222-9786 Supplier Research Products International Corp Supplier No. 400820
Popular HotPlate and Stirrer features a durable, chemically resistant white ceramic plate. Ideal space efficient design allows use on crowded benchtops or inside of hoods. Advanced microprocessor controls feature convenient turn knobs for quick, precise adjustment of speed and temperature.
LED light on front panel visually indicates when heat and/or stirring functions are activated. Work surface of 7 1/2 inches allows for a variety or laboratory vessels. Supplied with support rod for mounting thermometer or temperature probe.