Learn More
Hach Company Analog Insertion Mnt. ORP Sen.
Catalog No. 5000207284 Shop All Hach Company Products
Encompass
Hach Online Process ORP Sensor - General Purpose Analog Insertion Mount ORP SensorGeneral Purpose Online Process ORP Sensor - Platinum ORP Electrode PEEK Housing Insertion Mount 4 5m Cable Analog Cable Connection
Catalog No. 50-002-07284 Supplier Hach Company Supplier No. RD2P5
Hach Online Process ORP Sensor - General Purpose Analog Insertion Mount ORP SensorGeneral Purpose Online Process ORP Sensor - Platinum ORP Electrode PEEK Housing Insertion Mount 4 5m Cable Analog Cable Connection