Learn More
Hach Company Analog Insertion Mnt. ORP Sen.
Catalog No. 5000207312 Shop All Hach Company Products
Encompass
Hach Online Process ORP Sensor - General Purpose Analog Insertion Mount ORP SensorGeneral Purpose Online Process ORP Sensor - Platinum ORP Electrode PEEK Housing Insertion Mount 50 ft Cable Analog Cable Connection
Catalog No. 50-002-07312 Supplier Hach Company Supplier No. RD2P5A50
Hach Online Process ORP Sensor - General Purpose Analog Insertion Mount ORP SensorGeneral Purpose Online Process ORP Sensor - Platinum ORP Electrode PEEK Housing Insertion Mount 50 ft Cable Analog Cable Connection