Learn More
Hach Company Analog Jct. Box PVC Pipe-mount
Catalog No. 5000207276 Shop All Hach Company Products
Encompass
Analog Junction Box with PVC Pipe-mount (for 1-inch diameter pipe)Junction Box Pipe-Mount Pvc For 1-Inch Diameter Pipe Iincludes Mounting Hardware Only Used When The Desired Length Of Cable Between Analog Sensor And Analog Controller Is Greater Than Standard Length Of Sensor Cable
Catalog No. 50-002-07276 Supplier Hach Company Supplier No. 60G2052
Analog Junction Box with PVC Pipe-mount (for 1-inch diameter pipe)Junction Box Pipe-Mount Pvc For 1-Inch Diameter Pipe Iincludes Mounting Hardware Only Used When The Desired Length Of Cable Between Analog Sensor And Analog Controller Is Greater Than Standard Length Of Sensor Cable