Learn More
World Precision Instrument Analog Microforge-MF200-M1
SDP
Catalog No. 502521081
Encompass
The MF200 Microforge is a versatile instrument designed specifically for the fabrication of glass micropipettes and other related tools. The system was developed in collaboration with Dr. Ming Li of the Department of Pharmacology, University of South Alabama. The MF200 is simple, reliable and economically priced.
Catalog No. 50-252-1081 Supplier World Precision Instrument Supplier No. MF200M1
The MF200 Microforge is a versatile instrument designed specifically for the fabrication of glass micropipettes and other related tools. The system was developed in collaboration with Dr. Ming Li of the Department of Pharmacology, University of South Alabama. The MF200 is simple, reliable and economically priced.