Learn More
Julabo USA Inc Electronic module with analog connections for use with MAGIO MS and MX units.
SDP
Catalog No. NC1921808
Encompass_Preferred
Electronic module with analog connections for use with MAGIO™ MS and MX units. With one input and two outputs for external programming or flow sensor or temperature recorder (current/voltage), scalable as well as Standby input and Alarm output.
Catalog No. NC1921808 Supplier Julabo USA Inc Supplier No. 9900100
Electronic module with analog connections for use with MAGIO™ MS and MX units. With one input and two outputs for external programming or flow sensor or temperature recorder (current/voltage), scalable as well as Standby input and Alarm output.