Learn More
Hach Company Analog ORP Sensor PEEK
Catalog No. 5000207855 Shop All Hach Company Products
Encompass
Hach Online Process ORP Sensor - General Purpose Analog ORP Sensor-PEEK-4 5 m CableGeneral Purpose Online Process ORP Sensor - Platinum ORP Electrode PEEK Housing Convertible Mount 15 ft Cable Analog Cable Connection
Catalog No. 50-002-07855 Supplier Hach Company Supplier No. RD1P5A15V2S1N
Hach Online Process ORP Sensor - General Purpose Analog ORP Sensor-PEEK-4 5 m CableGeneral Purpose Online Process ORP Sensor - Platinum ORP Electrode PEEK Housing Convertible Mount 15 ft Cable Analog Cable Connection