Learn More
BINDER™ Analog Output
SDP
Catalog No. 502875723 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
KB 65/130/260/470/720/1060
KB PRO 130/260/470
KB PRO Series
KBF Series
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
50-287-5723 KB PRO Series
50-287-5601 KB 65/130/260/470/720/1060
50-287-5610 KB PRO 130/260/470
50-287-5721 KBF Series
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 50-287-5723 Supplier BINDER™ Supplier No. 80122375
Add to Cart
Add to Cart
Non-adjustable analog output
  • Analog output
  • Not adjustable
  • For temperature values or temperature and humidity values
  • 4 to 20 mA
Specifications
For Use With (Equipment) KB PRO Series
Material Copper
Type Analog Output
Features For Temperature and Humidity Values