Learn More
BINDER™ Analog Output 4-20 mA
SDP
Catalog No. 5000217492 Shop All BINDER Inc. Products
Click to view available options
Amperage:
4 to 20 mA
Catalog No. Amperage
50-002-17492 4 to 20 mA
1 options
Catalog No. 50-002-17492 Supplier BINDER™ Supplier No. 80121840
BINDER™ Analog Output 4-20 mA for use with CB 170 Carbon Dioxide Incubator.
  • For temperature and CO2 values with or without O2 control
  • Outputs not adjustable
Specifications
Description Analog output 4-20 mA - for temperature and CO2 values (outputs not adjustable), for versions with O2 control
For Use With (Equipment) CB 170
Type Analog Output
For Use With (Application) Temperature and Carbon Dioxide values with O2 control
Amperage 4 to 20 mA