Learn More
BINDER™ Analog Output 4-20 mA
SDP
Catalog No. 5000217493 Shop All BINDER Inc. Products
Click to view available options
For Use With (Equipment):
BINDER Chamber CB 170
Catalog No. For Use With (Equipment)
50-002-17493 BINDER Chamber CB 170
1 options
Catalog No. 50-002-17493 Supplier BINDER™ Supplier No. 80121839
BINDER™ Analog Output 4-20 mA for use with CB 170 Carbon Dioxide Incubator.
  • For temperature and CO2 values with or without O2 control
  • Outputs not adjustable
Specifications
Description Analog output 4-20 mA - for temperature and CO2 values (outputs not adjustable)
For Use With (Equipment) BINDER Chamber CB 170
Type Analog Output
For Use With (Application) Temperature and Carbon Dioxide values
Amperage 4 to 20 mA