Learn More
BINDER™ Analog Output
SDP
Catalog No. 08111121 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
08-111-121 Plastic
1 options
Catalog No. 08-111-121 Supplier BINDER™ Supplier No. 80121679
For Use With: BINDER units KMF 115
  • For temperature and humidity values
  • Output not adjustable
Specifications
Description Analog output 4-20 mA - for temperature and humidity values (output not adjustable)
For Use With (Equipment) BINDER Chambers KMF 115
Material Plastic
Type Analog Output
Amperage 4 to 20 mA