Learn More
Cole-Parmer Cole-Parmer Knick Instrinsically Safe Meter pH Electrode, Analog, Glass body electrode
Catalog No. NC1147283 Shop All Cole-Parmer Products
Encompass_Preferred
Optional analog probes for pH and conductivity measurement with intrinically-safe meters. Features a glass body electrode. Comes with a 1-m cable with connectors for Knick meters.
Catalog No. NC1147283 Supplier Cole-Parmer Supplier No. 5935122
Optional analog probes for pH and conductivity measurement with intrinically-safe meters. Features a glass body electrode. Comes with a 1-m cable with connectors for Knick meters.