Learn More
Vernier™ Analog Protoboard Adapter
SDPSCI_ED
Catalog No. S16103ND Shop All Vernier Software Products
Change view
Click to view available options
Type: Analog
Catalog No.
S16103ND Type: Analog
1 options
Catalog No. S16103ND Supplier Vernier™ Supplier No. BTAELV
Add to Cart
Add to Cart
Includes
Analog Protoboard Adapter
Requires
For use with standard prototyping boards or National Instruments ELVIS.
This connector makes it easy to wire the signal from a Vernier analog sensor into a non-Vernier interface.
  • The adapter fits into a standard prototyping board or National Instruments' ELVIS prototyping board, providing easy access to the sensor lines.
  • Vernier™ analog sensors require a 5 volt source and in most cases provide a 0 - 5 volt raw signal.
Warranty and Services
5-year limited warranty
Specifications
Voltage 5 V
For Use With (Application) Educational use only
Product Type Analog