Learn More
Hach Company Analog San Conduct Sens-CIP
Catalog No. 5000207448 Shop All Hach Company Products
Encompass
Analog Sanitary Flange Style Conductivity Sensor-Cell Constant 10 0-2 inch CIP FlangeNominal Cell Constant (K) 10 0 Sanitary (CIP) 2 inch Flange 6m (20 ft) Analog Cable 316 Stainless Steel Electrode
Catalog No. 50-002-07448 Supplier Hach Company Supplier No. 3455E7A
Analog Sanitary Flange Style Conductivity Sensor-Cell Constant 10 0-2 inch CIP FlangeNominal Cell Constant (K) 10 0 Sanitary (CIP) 2 inch Flange 6m (20 ft) Analog Cable 316 Stainless Steel Electrode