Learn More
Hach Company Analog San Conduct Sens-CIP
Catalog No. 5000207607 Shop All Hach Company Products
Encompass
Analog Sanitary Cconductivity Sensor-Cell Constant 10 0-2 inch CIP Flange-PP HDNominal Cell Constant (K) 10 0 Sanitary (CIP) 2 inch Flange Polypropylene Junction Box with 3/4in NPT Conduit Connection - Requires 1W1100 Interconnect Cable 316 Stainless Steel Electrode
Catalog No. 50-002-07607 Supplier Hach Company Supplier No. 3455E7B
Analog Sanitary Cconductivity Sensor-Cell Constant 10 0-2 inch CIP Flange-PP HDNominal Cell Constant (K) 10 0 Sanitary (CIP) 2 inch Flange Polypropylene Junction Box with 3/4in NPT Conduit Connection - Requires 1W1100 Interconnect Cable 316 Stainless Steel Electrode