Learn More
Braintree Scientific Analog Sensor Interface adapter 1 analog input (12V max)
SDP
Catalog No. 501954394
Encompass
Vary the pumping rate in proportion to a variable voltage input Start or stop pump at set voltages Vary the pumping rate to maintain a voltage window Attaches to any pump in the BS-Series Family **Sold for research purposes only and not for human use**
Catalog No. 50-195-4394 Supplier Braintree Scientific Supplier No. BSANABOX
Vary the pumping rate in proportion to a variable voltage input Start or stop pump at set voltages Vary the pumping rate to maintain a voltage window Attaches to any pump in the BS-Series Family **Sold for research purposes only and not for human use**