Learn More
Yamato Scientific America, Inc. Yamato 4L Constant Temperature Water Bath, Analog, 115V
Catalog No. 501898142 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Easy to use, compact design, 4L water bath. Analog set up system . Thermometer is included to verify actual temperature. Protected water tank prevents burns caused by contact. Temperature range: ambient +5~95°C.
Catalog No. 50-189-8142 Supplier Yamato Scientific America, Inc. Supplier No. BM100
Easy to use, compact design, 4L water bath. Analog set up system . Thermometer is included to verify actual temperature. Protected water tank prevents burns caused by contact. Temperature range: ambient +5~95°C.