Learn More
Cole-Parmer Analtech Uniplates, 20x20cm/UV/Soft/Silica G/250um Thick/15um Particle; 25/Pk
Catalog No. NC1920660 Shop All Cole-Parmer Products
Encompass
These TLC plates combine top-quality adsorbents, consistent manufacturing, rigid QC, and careful packaging--the results of 40 years of experience dedicated to thin layer techniques. Select the perfect analytical plates for your application. Choose plates with or without an acid stable fluorescent indicator (UV 254 nm). Scored plates snap apart evenly to facilitate single-use separations or screening.
Catalog No. NC1920660 Supplier Cole-Parmer Supplier No. 3410105
These TLC plates combine top-quality adsorbents, consistent manufacturing, rigid QC, and careful packaging--the results of 40 years of experience dedicated to thin layer techniques. Select the perfect analytical plates for your application. Choose plates with or without an acid stable fluorescent indicator (UV 254 nm). Scored plates snap apart evenly to facilitate single-use separations or screening.