Learn More
Miles Scientific Plate Holder (Plastic), Holds 25 plates
SDP
Catalog No. NC1559503
Encompass_Preferred
This TLC plate holder is lightweight and made of polypropylene. It is thus highly resistant to contamination or other ill affects from exposure to typical TLC solvents.The sloping bottom grooves guide TLC plates to the vertical grooves which hold plates in an upright position. Holds up to 25 TLC plates of any size. Dimensions: 10.5" x 8.0" x 4.0" (L x W x H)
Catalog No. NC1559503 Supplier Miles Scientific Supplier No. A5002
This TLC plate holder is lightweight and made of polypropylene. It is thus highly resistant to contamination or other ill affects from exposure to typical TLC solvents.The sloping bottom grooves guide TLC plates to the vertical grooves which hold plates in an upright position. Holds up to 25 TLC plates of any size. Dimensions: 10.5" x 8.0" x 4.0" (L x W x H)