Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Analysis of DNA Methylation using Restriction Enzymes Experiment Kit
SDPSCI_ED

Catalog No. S42875 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
6 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S42875 6 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S42875 Supplier Edvotek™ Supplier No. 205
Add to Cart
Add to Cart

Training kit to understand mechanisms of DNA methylation.

  • Explores effects of DNA methylation on restriction enzyme activity
  • Plasmid DNA will be digested with the restriction enzymes DpnI and DpnII
  • Methylated, unmethylated DNA will produce restriction fragments that are distinct from one another
  • Restriction fragments analyzed using agarose gel electrophoresis

Specifications

Grade 9 to Undergraduate
For Use With (Application) Educational purpose
Includes Instructions, methylated, unmethylated Plasmid DNA, EdvoQuick™ DNA Ladder, DpnI, DpnII restriction enzymes, agarose powder, InstaStain™ Ethidium Bromide, FlashBlue™ stain
Quantity 6 sets of restriction digestions
Time Range 2.5 hr.
Content And Storage Refrigeration required
Class Size 6 Lab Groups
Product Type Analysis of DNA Methylation using Restriction Enzymes Experiment Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.