Learn More
AIRGAS ANALYT CYLINDER REGULATOR 2STG
Catalog No. NC2207910
Encompass_Preferred
Airgas® Two Stage Brass 0-25 psi Analytical Cylinder Regulator With Needle Outlet CGA-580
This item is not returnable. View return policy
Catalog No. NC2207910 Supplier AIRGAS Supplier No. Y12244A580AG
Airgas® Two Stage Brass 0-25 psi Analytical Cylinder Regulator With Needle Outlet CGA-580