Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Free Acids Analyte Mix with Surrogate, SPEX CertiPrep™
SDP

Catalog No. 5522AS Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5522-AS 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5522-AS Supplier SPEX CertiPrep Supplier No. 5522AS

missing translation for 'validMainframeMsg'

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Tribromoacetic acid75-96-72000μg/mL
2,3-Dibromopropionic acid600-05-51000μg/mL
Chlorodibromoacetic acid5278-95-51000μg/mL
Chloroacetic acid79-11-8600μg/mL
Dichloroacetic acid79-43-6600μg/mL
Bromoacetic acid79-08-3400μg/mL
Bromochloroacetic acid5589-96-8400μg/mL
Bromodichloroacetic acid71133-14-7400μg/mL
Dalapon75-99-0400μg/mL
Dibromoacetic acid631-64-1200μg/mL
Trichloroacetic acid76-03-9200μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Free Acids Analyte Mix with Surrogate
Packaging 2mL amber prescored glass ampule
Name Note Method 552.2
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Methods For US EPA Method 552, 552.2
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solvent or Matrix MTBE
Quantity 1 mL

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.