Learn More
BUCHI Analytic PTFE Stopcock
SDP
Catalog No. 05405082 Shop All BUCHI Products
Change view
Click to view available options
Material:
PTFE
Catalog No. Material
05-405-082 PTFE
1 options
Catalog No. 05-405-082 Supplier BUCHI Supplier No. 11069607
Add to Cart
Add to Cart
For feeding of solvents and aerating a Rotavapor system with less cross-contamination compared to standard stopcocks. Ideal for applications where grease should be avoided or is undesired. Fits and replaces all stopcocks with 18.8/38 joint sizes.
  • No tubing included
Specifications
Material PTFE
Type Stopcock
For Use With (Application) Feeding of Solvents, and Aerating a Rotavapor System
For Use With (Equipment) R-100, R-300, R-210, R-215, RII
Standard Taper Joint Number Center 18.8/38