Learn More
Analytical Sales & Services Analytical Adjustable Flow Splitter, Pre-Column, Split Ratio Range - 5:1 to 100:1
Catalog No. 5000804753
Encompass
Analytical Adjustable Flow Splitter, Pre-Column\n\nSplit Ratio Range - 5:1 to 100:1\n\nQty/Pkg: EACH
Catalog No. 50-008-04753 Supplier Analytical Sales & Services Supplier No. 60104
Analytical Adjustable Flow Splitter, Pre-Column\n\nSplit Ratio Range - 5:1 to 100:1\n\nQty/Pkg: EACH