Learn More
Azzota Corp Analytical Balance 120g Ext Ca
Catalog No. 5000887241
Encompass_Preferred
Single weighting Analytical Balance 120g Capacity 0 0001g(0 1mg) Readability /-0 3mg Linearity Dia 80mm Pan External calibration windscreen included
Catalog No. 50-008-87241 Supplier Azzota Corp Supplier No. AUM120E
Single weighting Analytical Balance 120g Capacity 0 0001g(0 1mg) Readability /-0 3mg Linearity Dia 80mm Pan External calibration windscreen included