Learn More
Azzota Corp Analytical Balance 120g Int Ca
Catalog No. 5000887258
Encompass_Preferred
Single weighting Analytical Balance 120g Capacity 0 0001g(0 1mg) Readability /-0 3mg Linearity Dia 80mm Pan Internal calibration windscreen included
Catalog No. 50-008-87258 Supplier Azzota Corp Supplier No. AUM120I
Single weighting Analytical Balance 120g Capacity 0 0001g(0 1mg) Readability /-0 3mg Linearity Dia 80mm Pan Internal calibration windscreen included