Learn More
Azzota Corp Analytical Balance 150g Ext Ca
Catalog No. 5000887245
Encompass_Preferred
Single weighting Analytical Balance 150g Capacity 0 0001g(0 1mg) Readability /-0 3mg Linearity Dia 80mm Pan External calibration windscreen included
Catalog No. 50-008-87245 Supplier Azzota Corp Supplier No. AUM150E
Single weighting Analytical Balance 150g Capacity 0 0001g(0 1mg) Readability /-0 3mg Linearity Dia 80mm Pan External calibration windscreen included