Learn More
Azzota Corp Analytical Balance 210g Int Ca
Catalog No. 5000887233
Encompass_Preferred
Single weighting Analytical Balance 210g Capacity 0 0001g(0 1mg) Readability /-0 3mg Linearity Dia 80mm Pan Internal calibration windscreen included
Catalog No. 50-008-87233 Supplier Azzota Corp Supplier No. AUM210I
Single weighting Analytical Balance 210g Capacity 0 0001g(0 1mg) Readability /-0 3mg Linearity Dia 80mm Pan Internal calibration windscreen included