Learn More
Azzota Corp Analytical Balance 250g Ext Ca
Catalog No. 5000887247
Encompass_Preferred
Single weighting Analytical Balance 250g Capacity 0 0001g(0 1mg) Readability /-0 3mg Linearity Dia 80mm Pan external calibration windscreen included
Catalog No. 50-008-87247 Supplier Azzota Corp Supplier No. AUM250E
Single weighting Analytical Balance 250g Capacity 0 0001g(0 1mg) Readability /-0 3mg Linearity Dia 80mm Pan external calibration windscreen included