Learn More
Cole-Parmer Cole-Parmer Symmetry TA-314.C Analytical Balance with Touchscreen, 310g x 0.1mg, Internal Calibration
Catalog No. NC1688022 Shop All Cole-Parmer Products
Encompass_Preferred
The weighing surface of this balance allows for precise weighing of small items. Its user-friendly draft shield offers easy access to the weighing pan, while IR proximity touchless sensors offer touch-free balance control. It also has a color capacitive touch-screen display to provide clear visibility of weighing results. It has USB, RS-232, Ethernet, and Wi-Fi interfaces to offer secure data connections.
Catalog No. NC1688022 Supplier Cole-Parmer Supplier No. 1010203
The weighing surface of this balance allows for precise weighing of small items. Its user-friendly draft shield offers easy access to the weighing pan, while IR proximity touchless sensors offer touch-free balance control. It also has a color capacitive touch-screen display to provide clear visibility of weighing results. It has USB, RS-232, Ethernet, and Wi-Fi interfaces to offer secure data connections.