Learn More
Alkali Scientific Analytical Balance W3002A-120
SDP
Catalog No. 503029487
Encompass_Preferred
Introducing the Alkali Scientific Series Tx a perfect balance of performance design and quality Upgraded Weighing Performance Electromagnetic Force Restoration (EFR) technology fast response and stabilization and automatic internal calibration Refined Control Panels Enhanced color touch screen interface significantly improve ease of use Space Saving A careful redesign of our weigh cell assemblies has allowed us to pack more technology into a compact 8 x 15 inch footprint Convenient Connectivity Optional thermal printer or USB flash drive can be used for result documentation Semi Micro Class (W3002A-120) Readability of 0 00001 g up to 62 g and 0 0001 g up to 120 g Alkali ScientificAnalytical Balance W3002A-120 series Tx internal calibration Touch Screen 62gx0 00001g /120gx0 0001g is the most advanced in the Alkali Scientific line The large high resolution touch screen presents the user with an icon-based menu system and allows quick and intuitive access to a wide range of u
Catalog No. 50-302-9487 Supplier Alkali Scientific Supplier No. W3002A120
Introducing the Alkali Scientific Series Tx a perfect balance of performance design and quality Upgraded Weighing Performance Electromagnetic Force Restoration (EFR) technology fast response and stabilization and automatic internal calibration Refined Control Panels Enhanced color touch screen interface significantly improve ease of use Space Saving A careful redesign of our weigh cell assemblies has allowed us to pack more technology into a compact 8 x 15 inch footprint Convenient Connectivity Optional thermal printer or USB flash drive can be used for result documentation Semi Micro Class (W3002A-120) Readability of 0 00001 g up to 62 g and 0 0001 g up to 120 g Alkali ScientificAnalytical Balance W3002A-120 series Tx internal calibration Touch Screen 62gx0 00001g /120gx0 0001g is the most advanced in the Alkali Scientific line The large high resolution touch screen presents the user with an icon-based menu system and allows quick and intuitive access to a wide range of u