Learn More
Alkali Scientific Analytical Balance W3101A-120
SDP
Catalog No. 503029488
Encompass_Preferred
Analytical Balance W3101A-120 series Dx internal calibration Graphical Display 120gx0 0001g is robust workhorse instruments that utilize Electromagnetic Force Restoration (EFR) technology EFR is the gold standard for fast and reliable sub milligram mass determinations The waterproof and wear resistant front panel has been designed to hold up to spills and years of constant use A brightly lit dot matrix graphical display clearly shows weighing results and also enables an intuitive menu system thats easy to work with Useful features such as dynamic animal weighing piece counting percentage weighing and more are quickly accessible The front feet of the Dx balances are adjustable for proper leveling of the on a stable surface and a leveling bubble behind the weighing chamber confirms proper installation A Quik-Calcalibration system helps ensure that the balance is always calibrated and ready for use Simply press the CAL button to perform an automatic calibration using the internal
Catalog No. 50-302-9488 Supplier Alkali Scientific Supplier No. W3101A120
Analytical Balance W3101A-120 series Dx internal calibration Graphical Display 120gx0 0001g is robust workhorse instruments that utilize Electromagnetic Force Restoration (EFR) technology EFR is the gold standard for fast and reliable sub milligram mass determinations The waterproof and wear resistant front panel has been designed to hold up to spills and years of constant use A brightly lit dot matrix graphical display clearly shows weighing results and also enables an intuitive menu system thats easy to work with Useful features such as dynamic animal weighing piece counting percentage weighing and more are quickly accessible The front feet of the Dx balances are adjustable for proper leveling of the on a stable surface and a leveling bubble behind the weighing chamber confirms proper installation A Quik-Calcalibration system helps ensure that the balance is always calibrated and ready for use Simply press the CAL button to perform an automatic calibration using the internal