Learn More
Alkali Scientific Analytical Balance W3102A-220
SDP
Catalog No. 503029489
Encompass_Preferred
Introducing the Alkali Scientific Series Tx a perfect balance of performance design and quality Upgraded Weighing Performance Electromagnetic Force Restoration (EFR) technology fast response and stabilization and automatic internal calibration Refined Control Panels Enhanced color touch screen interface significantly improve ease of use Space Saving A careful redesign of our weigh cell assemblies has allowed us to pack more technology into a compact 8 x 15 inch footprint Convenient Connectivity Optional thermal printer or USB flash drive can be used for result documentation Semi Micro Class (W3002A-120) Readability of 0 00001 g up to 62 g and 0 0001 g up to 120 g Alkali ScientificAnalytical Balance W3102A-220 series Tx internal calibration Touch Screen 220gx0 0001g is the most advanced in the Alkali Scientific line The large high resolution touch screen presents the user with an icon-based menu system and allows quick and intuitive access to a wide range of useful function
Catalog No. 50-302-9489 Supplier Alkali Scientific Supplier No. W3102A220
Introducing the Alkali Scientific Series Tx a perfect balance of performance design and quality Upgraded Weighing Performance Electromagnetic Force Restoration (EFR) technology fast response and stabilization and automatic internal calibration Refined Control Panels Enhanced color touch screen interface significantly improve ease of use Space Saving A careful redesign of our weigh cell assemblies has allowed us to pack more technology into a compact 8 x 15 inch footprint Convenient Connectivity Optional thermal printer or USB flash drive can be used for result documentation Semi Micro Class (W3002A-120) Readability of 0 00001 g up to 62 g and 0 0001 g up to 120 g Alkali ScientificAnalytical Balance W3102A-220 series Tx internal calibration Touch Screen 220gx0 0001g is the most advanced in the Alkali Scientific line The large high resolution touch screen presents the user with an icon-based menu system and allows quick and intuitive access to a wide range of useful function