Learn More
Cole-Parmer IKA 2900001 Economical Analytical Mill, 115 VAC, 50/60 Hz
Supplier: Cole-Parmer 0430010
Rugged mill has a 420 stainless steel (SS) beater for grinding substances with a hardness rating of up to 6 Mohs — order an interchangeable blade and beater separately for enhanced versatility. Heavy-duty PTFE 80-mL chamber is included — order a 250-mL chamber separately. Indicator light and safety switch protect against overheating or improper closure.