Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ Analytical Precision UltraClass Weight, No Certificate

Catalog No. 02218097 Shop All Troemner Products
Change view
Click to view available options
Weight (Metric):
20 mg
30 mg
50 mg
300 mg
3 g
20 g
200 g
300 g
500 g
1 kg
2 kg
3 kg
10 kg
20 kg
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric)
02-218-097 20 kg
02-217-026 20 mg
02-217-027 30 mg
02-217-029 50 mg
02-217-030 300 mg
02-217-031 3 g
02-217-032 20 g
02-217-034 200 g
02-217-037 300 g
02-217-038 500 g
02-217-039 1 kg
02-217-040 2 kg
02-217-043 3 kg
02-218-095 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 02-218-097 Supplier Troemner™ Supplier No. 30390201
Add to Cart
Add to Cart
Request a Quote

Combine high precision with the advantage of two-piece construction with the most precise two-piece weights available with weight tolerances that equal ANSI/ASTM E617 Class 0 or OIML R 111 Class E2 tolerances.

  • Used in extremely stable environments
  • Two-piece construction with adjusting cavity
  • Supplied in a precision-fit, high quality polycarbonate case
TRUSTED_SUSTAINABILITY

Specifications

Product Type Two-Piece Analytical Weight
Certifications/Compliance No Certificate
Classification UltraClass
Material Stainless Steel
Weight (Metric) 20 kg
Density 7.95 g/cm3 at 20°C
Tolerance 25 mg
Length (Metric) 14.478 cm
Height (Metric) Case 20.2184 cm
For Use With (Application) Reference standard calibration
For Use With (Equipment) Calibration of high-precision analytical balances with readability as low as 0.01 mg
Includes Cylindrical Weight
Quantity Each
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.