Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ Analytical Precision UltraClass Weight Set, NVLAP Accredited Certificate

Catalog No. 02217054 Shop All Troemner Products
Change view
Click to view available options
Weight (Metric):
(2) 200 g to 1 g
1 to 200 g
100 mg
500 to 1 g
500 to 1 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric)
02-217-054 1 to 200 g
02-217-057 100 mg
02-217-051 (2) 200 g to 1 g
02-217-048 500 to 1 mg
02-217-047 500 to 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02-217-054 Supplier Troemner™ Supplier No. 30390153
Add to Cart
Add to Cart

Combine high precision with the advantage of two-piece construction with the most precise two-piece weights available with weight tolerances that equal ANSI/ASTM E617 Class 0 or OIML R 111 Class E2 tolerances.

  • Used in extremely stable environments
  • 1 to 500 mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
  • Weights 1 g and above are of two-piece construction
  • Weight sets 1 mg-500 g are provided in a high-quality polycarbonate set case
TRUSTED_SUSTAINABILITY

Specifications

Product Type 1 g and above: Two-piece, 500 mg and below: One-piece
Certifications/Compliance NVLAP Accredited Certificate
Classification UltraClass
Material Troemner Alloy 8
Case Material Polycarbonate
Weight (Metric) 1 to 200 g
Density 8.03 g/cm3 at 20°C
Length (Metric) 20.32 cm
Height (Metric) Case 8.89 cm
For Use With (Application) Reference standard calibration
For Use With (Equipment) Calibration of high-precision analytical balances with readability as low as 0.01 mg
Includes Cylindrical Weight: 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, Polycarbonate Case, Forceps, Brush
Quantity 10/Set
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.