Learn More
PCE AMERICAS INC Analytical balance scale with piece counting function. RS-232 interface
SDP
Catalog No. NC3654597
Encompass_Preferred
"The analytical balance scale is a scale for daily laboratory use. Another function besides the normal measurement function of the analytical balance scale is the piece counting function. This function allows the analytical balance scale to determine the number of screws, for example. Thus, for example, the analytical balance scale can also be used in logistics. So that the display is readable even in poor lighting conditions, the display of the analytical balance scale has a backlight. In addition to the measuring functions, the analytical balance scale also has an RS 232 interface. Thanks to this interface, the measured values ​​can be transferred to a PC, for example. This analytical balance scale can be used for stationary as well as for mobile use. For mobile use, the analytical balance scale requires 3 x 1.5V AA batteries, which can be used quickly and easily."
Catalog No. NC3654597 Supplier PCE AMERICAS INC Supplier No. PCEBSK 310
"The analytical balance scale is a scale for daily laboratory use. Another function besides the normal measurement function of the analytical balance scale is the piece counting function. This function allows the analytical balance scale to determine the number of screws, for example. Thus, for example, the analytical balance scale can also be used in logistics. So that the display is readable even in poor lighting conditions, the display of the analytical balance scale has a backlight. In addition to the measuring functions, the analytical balance scale also has an RS 232 interface. Thanks to this interface, the measured values ​​can be transferred to a PC, for example. This analytical balance scale can be used for stationary as well as for mobile use. For mobile use, the analytical balance scale requires 3 x 1.5V AA batteries, which can be used quickly and easily."