Learn More
Fluotics Analytik Jena Cybi Well compatible P10, sterile
Catalog No. 5000435123
Encompass_Preferred
Analytik Jena Cybi Well compatible P10, sterile, Working Volume: 10uL, Tip Color: Clear, Tip Tray, 384/25 uL, Compare To: OL3800-25-712-P, 24 racks/case with 384 tips/rack
Catalog No. 50-004-35123 Supplier Fluotics Supplier No. CYBW10.ST
Analytik Jena Cybi Well compatible P10, sterile, Working Volume: 10uL, Tip Color: Clear, Tip Tray, 384/25 uL, Compare To: OL3800-25-712-P, 24 racks/case with 384 tips/rack