Learn More
Fluotics Analytik Jena Cybi Well compatible P60, filter/sterile
Catalog No. 5000435275
Encompass_Preferred
Analytik Jena Cybi Well compatible P60, filter/sterile, Working Volume: 35uL, Tip Color: Clear, Tip Tray, 384/60 uL, Compare To: OL3800-25-915-F, 24 racks/case with 384 tips per rack
Catalog No. 50-004-35275 Supplier Fluotics Supplier No. CYBW60.F
Analytik Jena Cybi Well compatible P60, filter/sterile, Working Volume: 35uL, Tip Color: Clear, Tip Tray, 384/60 uL, Compare To: OL3800-25-915-F, 24 racks/case with 384 tips per rack